The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
- pCAL vectors derived from T7 RNA polymerase-based pET vectors
- Novel purification tag combines small size, gentle elution and high-affinity binding
- Easy tag removal resulting in native protein
: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
: For research use only. Not for use in diagnostic procedures.
- Description:pCAL-n-EK Vector
- Size:20 μg
- Details:Contains enterokinase cleavage site. Derived from pET-11a. Recombinant proteins expressed as N-terminal fusion to the CBP affinity tag.