Order Entry
United States
Orders LinkContactUsLinkComponent
VWR® Buffer Solutions

VWR® Buffer Solutions

Phosphate, Rigid pack, 20 L, 7.00, Green
Catalog # CA71006-780
VWR® Buffer Solutions
Catalog # CA71006-780
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions

Phosphate, Rigid pack, 20 L, 7.00, Green

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER PH7 CALIBRATING 5GAL Catalog #RC1551-5

Product RC1551-5 is similar to CA71006-780 but manufacturer and specifications may vary.

BDH BUFFER PH 7 YELLOW 20L Catalog #BDH0194-20L

Product BDH0194-20L is similar to CA71006-780 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These buffer solutions are for calibrating pH meters

They are traceable to NIST standards.

Specifications

Specifications

  • Pack type:
    Rigid pack
  • Color:
    Green
  • Size:
    20 L
  • pH buffer type:
    Phosphate
  • pH:
    7.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.