Orders LinkContactUsLinkComponent
TechniSat® Presaturated Wipers, Texwipe®
TechniSat® Presaturated Wipers, Texwipe®
23×28 cm (9×11"), Non sterile, 70/reclosable bag
Catalog # TW-TX1041
Supplier:   Texwipe
Supplier #:  TX1041
TechniSat® Presaturated Wipers, Texwipe®
Catalog # TW-TX1041
Supplier:  Texwipe
TechniSat® Presaturated Wipers, Texwipe®
TechniSat® Presaturated Wipers, Texwipe®
23×28 cm (9×11"), Non sterile, 70/reclosable bag
Price
$63.10
New Customer Promotional Price
Log in for availability
Each Case of 12
Product Details & Documents
These convenient wipers provide portion control and reduced potential of spillage in and out of the cleanroom.
  • Fibers are blended by a hydroentanglement procedure
  • Uses water instead of chemicals to bind
  • Excellent bidirectional strength and a very low extractable level
  • Wipers are controlled for traceability
The blended material of 55% cellulose and 45% polyester provides the high absorbency of a natural fiber with the cleanliness and strength of a synthetic material. For non sterile wipers, the 70% semiconductor-grade isopropyl alcohol is filtered to 0.2 µm and diluted to 70% concentration with 18 megohm-cm deionized water. Nonsterile wipers are available in FlexPacks or recloseable bags. Sterile wipers are saturated with USP grade isopropyl alcohol and packaged in flexpacks. The flexpacks are then double-bagged to maintain sterility.
Specifications
  • Sterility:
    Non sterile
  • Packaging:
    70/reclosable bag
  • W×L:
    23×28 cm (9×11")