Order Entry
United States
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

5×20 μg
Catalog # PAV5113
Supplier:   Promega Corporation
Supplier #:  V5113
Trypsin
Catalog # PAV5113
CAS Number:  
Trypsin
Trypsin

5×20 μg

CAS Number:  
Price
$60.36
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequencing Grade Trypsin has been manufactured to provide maximum specificity. Lysine residues in porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is extremely resistant to autolytic digestion.

  • Manufactured for Maximum Specificity and Stability
  • Lysine residues modified by reductive methylation, yielding a highly active and stable trypsin
  • Available lyophilized or frozen
  • Referenced in thousands of papers

Trypsin is a serine protease that specifically cleaves at the carboxylic side of lysine and arginine residues. The stringent specificity of trypsin is essential for protein identification. Native trypsin is subject to autolysis, generating pseudotrypsin, which exhibits a broadened specificity including a chymotrypsin-like activity. Such autolysis products, present in a trypsin preparation, would result in additional peptide fragments that could interfere with database analysis of the mass of fragments detected by mass spectrometry. Sequencing Grade Trypsin has been manufactured to provide maximum specificity. Lysine residues in the porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is extremely resistant to autolytic digestion. The specificity of the purified trypsin is further improved by TPCK treatment, which inactivates chymotrypsin. The treated trypsin is then purified by affinity chromatography and lyophilized. It is resistant to mild denaturing conditions such as 0.1% SDS, 1 M urea or 10% acetonitrile and retains 50% of its activity in 2 M guanidine HCl. The activity of trypsin is decreased when acidic residues are present on either side of a susceptible bond. If proline is at the carboxylic side of lysine or arginine, the bond is almost completely resistant to cleavage. Recommended Reaction Buffer: 50 mM NH(4)HCO(3) (pH 7.8).

Documents

Specifications

Specifications

  • Size:
    5×20 μg
  • Storage Conditions:
    Store at –65 °C
  • Cat. No.:
    PAV5113
  • Form:
    Frozen
  • Grade:
    Sequencing grade
  • Enzyme Name:
    Trypsin

Specifications