Order Entry
United States
Orders LinkContactUsLinkComponent
XhoI

XhoI

XhoI, 3000 U
Catalog # PAR6161
Supplier:   Promega Corporation
Supplier #:  R6161
XhoI
Catalog # PAR6161
CAS Number:  
XhoI
XhoI

XhoI, 3000 U

CAS Number:  
Price
$62.68
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

C/TCGA G G AGCT\C

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications

Documents

Specifications

Specifications

  • Description:
    XhoI
  • Size:
    3000 U
  • Storage Conditions:
    –30...–10 °C
  • Cat. No.:
    PAR6161
  • Buffer supplied with enzyme:
    Buffer D
  • Enzyme assay temperature:
    37°C
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 10…25%
  • Enzyme Name:
    XhoI

Specifications