Order Entry
United States
Orders LinkContactUsLinkComponent
PstI

PstI

PstI, 15000 U
Catalog # PAR6115
Supplier:   Promega Corporation
Supplier #:  R6115
PstI
Catalog # PAR6115
CAS Number:  
PstI
PstI

PstI, 15000 U

CAS Number:  
Price
$182.99
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

C TGCA/G G\ACGT C

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, provides a higher level of quality control for enzymes used in cloning applications

Documents

Specifications

Specifications

  • Description:
    PstI
  • Size:
    15000 U
  • Storage Conditions:
    –30...–10 °C
  • Cat. No.:
    PAR6115
  • Buffer supplied with enzyme:
    Buffer H
  • Enzyme assay temperature:
    37°C
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
  • Enzyme Name:
    PstI

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.