Order Entry
United States
Orders LinkContactUsLinkComponent
 

PstI

PstI, 3000 U
Catalog # PAR6111
Supplier:   Promega Corporation
 :  R6111
PstI
Catalog # PAR6111
CAS Number:  
PstI
PstI

PstI, 3000 U

CAS Number:  
Price
$50.55
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

C TGCA/G G\ACGT C

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, provides a higher level of quality control for enzymes used in cloning applications

 

Specifications

Specifications

  • Description:
    PstI
  • Size:
    3000 U
  • Storage Conditions:
    –30...–10 °C
  • Cat. No.:
    PAR6111
  • Buffer supplied with enzyme:
    Buffer H
  • Enzyme assay temperature:
    37°C
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
  • Enzyme Name:
    PstI

Specifications