Orders LinkContactUsLinkComponent
Affinity Protein Expression and Purification Systems, Agilent Technologies
Affinity Protein Expression and Purification Systems, Agilent Technologies
pCAL-c Vector, Derived from pET-11d. Recombinant proteins expressed as C-terminal fusion to the CBP affinity tag. Thrombin-cleavage site is immediately downstream of BamH I cloning site., 20 μg
Catalog # 99900-884
Affinity Protein Expression and Purification Systems, Agilent Technologies
Catalog # 99900-884
Supplier:  
CAS Number:  
Affinity Protein Expression and Purification Systems, Agilent Technologies
Affinity Protein Expression and Purification Systems, Agilent Technologies
pCAL-c Vector, Derived from pET-11d. Recombinant proteins expressed as C-terminal fusion to the CBP affinity tag. Thrombin-cleavage site is immediately downstream of BamH I cloning site., 20 μg
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
  • pCAL vectors derived from T7 RNA polymerase-based pET vectors
  • Novel purification tag combines small size, gentle elution and high-affinity binding
  • Easy tag removal resulting in native protein
Delivery information: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Caution: For research use only. Not for use in diagnostic procedures.
Documents
Specifications
Specifications
  • Description:
    pCAL-c Vector
  • Size:
    20 μg
  • Details:
    Derived from pET-11d. Recombinant proteins expressed as C-terminal fusion to the CBP affinity tag. Thrombin-cleavage site is immediately downstream of BamH I cloning site.