Orders LinkContactUsLinkComponent
pBlueScript II Vectors, Agilent Technologies
pBlueScript II Vectors, Agilent Technologies
R408 Interference Resistant Helper Phage, 1 ml, Stable interference-resistant helper phage (4 kB, single stranded). For preferential secretion of single-stranded vectors containing intact segments of an M13 intergenic region over the helper phage DNA
Catalog # 95040-870
pBlueScript II Vectors, Agilent Technologies
Catalog # 95040-870
Supplier:  
CAS Number:  
pBlueScript II Vectors, Agilent Technologies
pBlueScript II Vectors, Agilent Technologies
R408 Interference Resistant Helper Phage, 1 ml, Stable interference-resistant helper phage (4 kB, single stranded). For preferential secretion of single-stranded vectors containing intact segments of an M13 intergenic region over the helper phage DNA
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
  • High copy number ColE1-based phagemid
  • Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
  • T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Description:
    R408 Interference Resistant Helper Phage
  • Size:
    1 ml
  • Details:
    Stable interference-resistant helper phage (4 kB, single stranded). For preferential secretion of single-stranded vectors containing intact segments of an M13 intergenic region over the helper phage DNA