Order Entry
United States
Orders LinkContactUsLinkComponent
Collodion USP

Collodion USP

Glass, amber, 100 ml
Catalog # 700012-952
Flammable
Irritant
Supplier #:  CO120-100MLG
Collodion USP
Catalog # 700012-952
CAS Number:  9004-70-0
Collodion USP

Glass, amber, 100 ml

CAS Number:  9004-70-0
Price
$196.00
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

  • CAS Number:9004-70-0
  • MDL Number:MFCD00081525
  • Merck Index:13,02508
  • Boiling Pt:∼34 °C (1013 hPa)
  • Melting Pt:160 °C
  • Density:0.75 g/cm³ (20 °C)
  • Flash Pt:–19 °C
  • Storage Temperature:Ambient
  • UN:2059
  • ADR:3,II

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Assay:
    5% Min.

Specifications

  • Pack type:
    Glass, amber
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.