Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dehydrant Blends, Epredia

Dehydrant 70%, Poly Bottle, 1 gal.
Catalog # 66004-948
Supplier:   Epredia
 :  6501
Dehydrant Blends, Epredia
Catalog # 66004-948
Supplier:  Epredia
CAS Number:  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia

Dehydrant 70%, Poly Bottle, 1 gal.

CAS Number:  
Price
$306.59
New Customer Promotional Price
Log in for availability
Case of 4 (950ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.

  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents

Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.

Specifications

Specifications

  • Pack type:
    Poly Bottle
  • Description:
    Dehydrant 70%
  • Size:
    1 gal.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.