Order Entry
United States
Orders LinkContactUsLinkComponent
epT.I.P.S.® Racks - Pipette Tips

epT.I.P.S.® Racks - Pipette Tips

50 - 1250 µl, 76 mm, Green, 960 Tips (10 Racks×96 Tips)
Catalog # 47751-667
Supplier:   Eppendorf
Supplier #:  0030071590
epT.I.P.S.® Racks - Pipette Tips
Catalog # 47751-667
Supplier:  Eppendorf
epT.I.P.S.® Racks - Pipette Tips
epT.I.P.S.® Racks - Pipette Tips

50 - 1250 µl, 76 mm, Green, 960 Tips (10 Racks×96 Tips)

Price
$179.00
Log in for availability
Case of 960
Minimum order quantity is 1
Product Details & Documents

Eppendorf epT.I.P.S.® Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur®', and 'Sterile' purity grades. All tips are supplied in disposible racks designed for optimal organization and user convenience in demanding laboratory environments.

  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.

Certifications: Lot-specific certficates available for the purity grades PCR clean, Sterile, and Biopur® can be downloaded from the Eppendorf website.

Documents

Specifications
  • Volume:
    50 - 1250 µl
  • Length:
    76 mm
  • Packed:
    960 Tips (10 Racks×96 Tips)
  • Color code:
    Green
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.