Order Entry
United States
Orders LinkContactUsLinkComponent
Corning® Trypsin

Corning® Trypsin

0.25%Trypsin, 2.21 mM EDTA, 1X [-] sodium bicarbonate, 100 ml
Catalog # 45000-664
Supplier:   Corning
Supplier #:  25-053-CI
Corning® Trypsin
Catalog # 45000-664
Supplier:  Corning
CAS Number:  
Corning® Trypsin
Corning® Trypsin

0.25%Trypsin, 2.21 mM EDTA, 1X [-] sodium bicarbonate, 100 ml

CAS Number:  
Price
$97.40
Log in for availability
Pack of 6 (100ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.

  • Storage: -25°C to -15°C

Trypsin has a pH specification range from 7.2 to 8. At this range, phenol red inclusion gives the solution a pink color. During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not affect the functionality of the trypsin. By adjusting the pH to 7.2 – 8 with NaOH the trypsin activity could be restored.

Specifications

Specifications

  • Description:
    0.25%Trypsin, 2.21 mM EDTA, 1X [-] sodium bicarbonate
  • Size:
    100 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.