Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dulbecco's phosphate-buffered saline (DPBS), Mediatech

DPBS, Corning® 1 L DPBS (Dulbeccos Phosphate Buffered Saline), 1X [-] calcium, magnesium, 1 L, +2 °C…+30 °C
  45000-436
 :   Corning
 :  21-031-CM
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
  45000-436
 :  Corning
 :  
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Dulbecco's phosphate-buffered saline (DPBS), Mediatech

DPBS, Corning® 1 L DPBS (Dulbeccos Phosphate Buffered Saline), 1X [-] calcium, magnesium, 1 L, +2 °C…+30 °C

 :  
 
$240.00
   
Pack of 6 (1l)
 

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

Complete Certificate of Analysis available for each production lot, along with full formulation information.

It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.

 : Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C

 

 

  • Description:
    DPBS, Corning® 1 L DPBS (Dulbeccos Phosphate Buffered Saline), 1X [-] calcium, magnesium
  • Storage temperature:
    +2 °C…+30 °C
  • Volume:
    1 L
  • Cat. No.:
    45000-436
  • Storage Temperature:
    Ambient

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.