Order Entry
United States
Orders LinkContactUsLinkComponent
VWR® Corks

VWR® Corks

38 mm, 22, 38 mm, 44 mm
Catalog # 23420-480
   Low Manufacturing Impact
VWR® Corks
Catalog # 23420-480
Supplier:  
   Low Manufacturing Impact
VWR® Corks
VWR® Corks

38 mm, 22, 38 mm, 44 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VWR CORKS REG PK100 N0.5 Catalog #23420-162

Product 23420-162 is a replacement for 23420-480 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

XXXX Corks function as a closure for bottles, vials, laboratory and other openings.

  • Regular length taper
  • Assorted sizes

The XXXX premium grade corks have fewer lenticels (crevices) than standard grade

Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.

This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.

Documents

Specifications
  • Height:
    38 mm
  • Stopper no.:
    22
  • Top diameter:
    44 mm
  • Bottom diameter:
    38 mm
  • Cat. No.:
    23420-480
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.