Orders LinkContactUsLinkComponent
VWR® Corks
VWR® Corks
Assorted, Nos. 0 to 11
Catalog # 23416-008
   Low Manufacturing Impact
Supplier:   VWR International
VWR® Corks
Catalog # 23416-008
Supplier:  VWR International
   Low Manufacturing Impact
VWR® Corks
VWR® Corks
Assorted, Nos. 0 to 11
Price
$67.70
New Customer Promotional Price
Log in for availability
Pack of 100
Product Details & Documents
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
  • Regular length taper
  • Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade. This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.
Specifications
  • Description:
    Assorted, Nos. 0 to 11
  • Cat. No.:
    23416-008
  • Environmentally Preferable:
    Low Manufacturing Impact