solution 100 mg/mL
Product 101414-060 is a replacement for 100218-132 and has the same packaging and specifications, but may be a different quantity.
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.
It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.
- CAS Number:4697-36-3
- Formula:C₁₇H₁₈N₂O₆S
Documents
Specifications
Specifications
- Size:25 ml
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



