Order Entry
United States
Orders LinkContactUsLinkComponent
BBL™ Phosphate Buffer, BD

BBL™ Phosphate Buffer, BD

Phosphate buffer, 99 ml, 7.2
Catalog # 90004-214
Supplier:   BD
Supplier #:  292328
BBL™ Phosphate Buffer, BD
Catalog # 90004-214
Supplier:  BD
CAS Number:  
BBL™ Phosphate Buffer, BD

Phosphate buffer, 99 ml, 7.2

CAS Number:  
Price
$327.69
New Customer Promotional Price
Log in for availability
Pack of 10 (99ml)
Minimum order quantity is 1
Product Details & Documents

This is used as buffered diluent for microbiological procedures.

Specifications

Specifications

  • Description:
    Phosphate buffer
  • Size:
    99 ml
  • pH:
    7.2
  • Cat. No.:
    90004-214
  • Shelf Life:
    365

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.