Order Entry
United States
Orders LinkContactUsLinkComponent
 

Difco™ m Enterococcus Agar, BD Diagnostics

m Enterococcus Agar, 500 g
Catalog # 90003-932
Supplier:   BD
 :  274620
Difco™ m Enterococcus Agar, BD Diagnostics
Catalog # 90003-932
Supplier:  BD
CAS Number:  
Difco™ m Enterococcus Agar, BD Diagnostics

m Enterococcus Agar, 500 g

CAS Number:  
Price
$438.57
New Customer Promotional Price
Log in for availability
Each (500g)
Product Details & Documents

Selective isolation and enumeration of enterococci using the membrane filtration technique.

 

Specifications

Specifications

  • Description:
    m Enterococcus Agar
  • Size:
    500 g

Specifications