Orders LinkContactUsLinkComponent
SICKLEDEX® Solubility Test Kit, Streck
SICKLEDEX® Solubility Test Kit, Streck, 100
Catalog # 87005-414
Supplier:   Streck Laboratories
 :  217657
SICKLEDEX® Solubility Test Kit, Streck
Catalog # 87005-414
CAS Number:  
SICKLEDEX® Solubility Test Kit, Streck
SICKLEDEX® Solubility Test Kit, Streck
SICKLEDEX® Solubility Test Kit, Streck, 100
CAS Number:  
Price
$519.03
Log in for availability
Box of 2
Product Details & Documents
Qualitative solubility kits are used to detect the presence of sickling hemoglobins in human blood or sickle cell control material
A ten-place test tube reading rack is also available.
The pipet-style tests produce results in just six minutes. The kits are conveniently packaged with two separate squeeze bottles of phosphate buffer with Saponin for easy dispensing, and to eliminate waste. The kits also ontain two buffer dispensing caps and two vials of sodium hydrosulfite reagent power. They have 45-day open-vial stability and one-year closed-vial stability.
Specifications
Specifications
  • Description:
    SICKLEDEX® Solubility Test Kit, Streck
  • No. of tests:
    100
Specifications
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Test Tube Rack
    Test Tube Rack
    87005-416
    Log in for availability
    Each - $56.06