Orders LinkContactUsLinkComponent
Para 4® Controls, Streck
High Level, 1.5 mL
Catalog # 87005-408
Supplier:   Streck Laboratories
 :  215436
Para 4® Controls, Streck
Catalog # 87005-408
CAS Number:  
Para 4® Controls, Streck
Para 4® Controls, Streck
High Level, 1.5 mL
CAS Number:  
Price
$99.91
Log in for availability
Box of 6
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
These four-parameter hematology controls are designed for laboratories that perform a limited number of hematology tests
The assay includes Hgb, Hct, and platelet values. It is also assayed for auxiliary methods. Controls have an open-vial stability for 14 days and a closed-vial stability for 110 days. Available in glass or plastic vials.
Specifications
Specifications
  • Description:
    High Level
  • Size:
    1.5 mL
Specifications