Order Entry
United States
Orders LinkContactUsLinkComponent
Para 12® Plus Controls, Streck

Para 12® Plus Controls, Streck

Normal Level, 3.0 mL
Catalog # 87004-510
Supplier:   Streck Laboratories
Supplier #:  218914
Para 12® Plus Controls, Streck
Catalog # 87004-510
CAS Number:  
Para 12® Plus Controls, Streck
Para 12® Plus Controls, Streck

Normal Level, 3.0 mL

CAS Number:  
Price
$209.02
Log in for availability
Pack of 4 (3ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A hematology control with assay values provided for the five-part white cell differential

Open-vial stability is seven days and closed-vial stability is 75 days. Packaged in a vial with pierceable cap. Compatible with Abbott® instruments.

Specifications

Specifications

  • Description:
    Normal Level
  • Size:
    3.0 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.