Orders LinkContactUsLinkComponent
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Peach
  87002-486
 :  M498-7
Histosette® I Tissue Processing/Embedding Cassettes, Simport
  87002-486
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Peach
$414.58
Case of 1,500
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids snap apart and lock easily into the base of the cassettes. The anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments.
 : Packed in three dispenser boxes of 500 cassettes. Fluorescent colored cassettes require a minimum order quantity of 40 cases for purchase.
  • Color:
    Peach