Order Entry
United States
Orders LinkContactUsLinkComponent
EasyDip™ Slide Staining System, Simport Scientific

EasyDip™ Slide Staining System, Simport Scientific

EasyDip™ Slide Staining Jars, 64L x 76W x 92H mm (21/2 x 3 x 35/8"), Pink
Catalog # 87000-124
Supplier:   Simport Scientific
Supplier #:  M900-12P
EasyDip™ Slide Staining System, Simport Scientific
Catalog # 87000-124
Supplier:  Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific

EasyDip™ Slide Staining Jars, 64L x 76W x 92H mm (21/2 x 3 x 35/8"), Pink

Price
$193.70
Log in for availability
Case of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The slide staining system is made of acetal plastic.

  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide stable base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications

The staining jar is made of resistant acetal plastic, and will not break like most glass jars do. It will resist most staining agents including alcohol and xylene (but not phenol, iodine, or ferric chloride). The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80 ml. Easy to clean and no metals to corrode. Ideal for special stains, frozen sections, and special processes.

The system is not autoclavable. Volume is 80 ml. System dimensions are 64×76×92 mm (2.5×3×3.625").

Ordering information: The staining jars and staining rack are sold separately. The EasyDip™ Kit with Rack includes one anodized aluminum holder along with six assorted color jars (two white ones) and one M905-12DGY Slide Staining Rack. The EasyDip™ Aluminum Holder is designed to hold six EasyDip™ slide staining jars. The assorted kit includes one jar of each color.

Specifications
  • Color:
    Pink
  • Description:
    EasyDip™ Slide Staining Jars
  • Dimensions:
    64L x 76W x 92H mm (21/2 x 3 x 35/8")

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.