Orders LinkContactUsLinkComponent
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Pink
Catalog # 82028-746
Supplier:   Simport Scientific
Supplier #:  M509-3
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Catalog # 82028-746
Supplier:  Simport Scientific
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Pink
Price
$533.65
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These compact, easy-to-use cassettes incorporate a unique recessed cover which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1x 5mm slotted openings ensure efficient fluid exchange and drainage, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle
Certifications: CE marked.
Ordering information: Packaged in three dispenser boxes of 500 cassettes with covers assembled.
Documents
Specifications
  • Color:
    Pink