BD Microtainer® Tubes, BD Diagnostics
Product Details & Documents
BD Microtainer® tubes are designed for the collection, transport, and processing of skin puncture blood.
The “twist-assist” BD Microgard™ closure is ready-to-use and reduces blood splatter upon removal.
The integrated collection scoop, along with a wider diameter tube, allow for ease of collection and improved mixing. To simplify additive and sample identification, Microtainer® tubes are color-coded in a manner similar to evacuated tubes. Volume level lines, printed on tubes, ensure proper blood-to-anticoagulant ratio.
Documents
Specifications
- Description:BD Microtainer® tube
- Additive:K2 EDTA
- Cap color:Lavender
Accessories
Product Information
- Catalog #AvailabilityPrice
- Tube Extender for use with VT365974