QuEChERS SPE Tubes, Restek
Graphite Carbon, 600 mg, 200 mg Graphitized Carbon and 400 mg PSA
RK26127
: Restek
: 26127
These tubes comply with the USDA's QuEChERS method, used in the extraction and clean-up of pesticide residue samples.
- Contained bed weight varies between models
- Adsorbants and secondary amine exchange materials differ between models
Tubes contain ultrapure adsorbants and primary and secondary amine exchange material (PSA). Fluoropolymer resin frits minimize background.
- Bed weight:600 mg
- Packing material:200 mg Graphitized Carbon and 400 mg PSA
- Phase:Graphite Carbon