QuEChERS SPE Tubes, Restek
Graphite Carbon, 250 mg, 150 mg Magensium Sulfate, 50 mg PSA, and 50 mg Graphitized Carbon
Catalog # RK26123
Supplier: Restek
Supplier #: 26123
Jump to:
Product Details & Documents
These tubes comply with the USDA's QuEChERS method, used in the extraction and clean-up of pesticide residue samples.
- Contained bed weight varies between models
- Adsorbants and secondary amine exchange materials differ between models
Tubes contain ultrapure adsorbants and primary and secondary amine exchange material (PSA). Fluoropolymer resin frits minimize background.
Specifications
- Bed weight:250 mg
- Packing material:150 mg Magensium Sulfate, 50 mg PSA, and 50 mg Graphitized Carbon
- Phase:Graphite Carbon