L(+)-Histidine USP, Multi-Compendial
Product MK4935-06 is similar to JT2080-5 but manufacturer and specifications may vary.
Product 95033-966 is similar to JT2080-5 but manufacturer and specifications may vary.
Product 95033-960 is similar to JT2080-5 but manufacturer and specifications may vary.
Product Details & Documents
White Crystalline solid.
- CAS Number:71-00-1
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00064315
- Molecular Weight:155.16 g/mol
- Merck Index:13,04741
- Melting Pt:272…273 °C
- Density:1.44 g/cm³ (20 °C)
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- EINECS:200-745-3
Documents
Certificates
Search by Lot Number2080 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
JTBaker_2080_Datasheet.pdf
Datasheet
spec_JT2080-09 spec_JT2080-5 spec_JT2080-6 spec_JT2080-7
Product specification
Specifications
Specification Test Results
- USP - Assay (C₆H₉N₃O₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):12.6 - 14.0 Degree
- USP - pH:7.0 - 8.5
- USP - Loss on Drying at 105°C:≤ 0.2 %
- USP - Residue on Ignition:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds - Individual Impurities:≤ 0.5 %
- USP - Related Compounds - Total Impurities:≤ 2.0 %
- EP/BP - Assay (C₆H₉N₃O₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):11.4 - 12.4 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 0.5 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:≤ 0.5 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- JP - Assay (C6H9N3O2) (dried basis):99.0 - 101.0 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Chloride (Cl):≤ 0.021 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Iron (Fe):≤ 10 ppm
- JP - Identification:Passes Test
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP- Optical Rotation [Å]²º^D (+):11.8 - 12.8 Degree
- JP - Related Substances:Passes Test
- JP - pH:7.0 - 8.5
- JP - Residue on Ignition:≤ 0.10 %
- JP - Sulfate (SO₄):≤ 0.028 %
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



