Order Entry
United States
Orders LinkContactUsLinkComponent
3-(3-Fluorophenyl)propionic acid 97%

3-(3-Fluorophenyl)propionic acid 97%

bottle, 1 g
  AAH27847-03
 
Supplier #:  H27847-03
3-(3-Fluorophenyl)propionic acid 97%
  AAH27847-03
 :  458-45-7
3-(3-Fluorophenyl)propionic acid 97%

bottle, 1 g

 :  458-45-7
 
$21.23
   
Each (1g)
 
   
 

Purity: 97% CAS No.: 458-45-7 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 110°(230°F) Melting Point: 43-47° Boiling Point: 125°/10mm MDL No.: MFCD01310821

 :  3-(3-Fluorophenyl)propanoic acid
  •  :458-45-7
  •  :C₉H₉FO₂
  •  :MFCD01310821
  •  :168.17
  •  :125 °C/10 mm
  •  :43…47 °C
  •  :110 °C(230 °F)
  •  :Ambient
 

 

  • Pack type:
    bottle
  • Size:
    1 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.