Order Entry
United States
Orders LinkContactUsLinkComponent
 

Polysorbate 80

Bottle, 500 ml
Catalog # AAAL13315-AP
 :  L13315-AP
Polysorbate 80
Catalog # AAAL13315-AP
CAS Number:  9005-65-6
Polysorbate 80

Bottle, 500 ml

CAS Number:  9005-65-6
Price
$79.31
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Used for solubilizing membrane proteins during isolation and purification

CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:14,7582
  • Boiling Pt:>100 °C
  • Melting Pt:100 °C
  • Density:1.064
  • Flash Pt:>149 °C(300 °F)
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.