Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dexamethasone 98%

Bottle, 5 g
Catalog # AAA17590-06
Health
 :  A17590-06
Dexamethasone 98%
Catalog # AAA17590-06
CAS Number:  50-02-2
Dexamethasone 98%
Dexamethasone 98%

Bottle, 5 g

CAS Number:  50-02-2
Price
$612.29
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Anti-inflammatory glucocorticoid

Purity: 98% CAS No.: 50-02-2 Molecular Formula: C22H29FO5 Formula Weight: 392.47 Melting Point: ca 260° dec. Beilstein Reference: 2066651 Merck Reference: 14,2943 Optical Rotation: +75° (c=1 in doxane) MDL No.: MFCD00064136

Anti-inflammatory glucocorticoid

Synonyms:  Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
  • CAS Number:50-02-2
  • Formula:C₂₂H₂₉FO₅
  • MDL Number:MFCD00064136
  • Molecular Weight:392.47 g/mol
  • Merck Index:14,2943
  • Melting Pt:∼ 260 °C (dec.)
  • Melting Pt:ca 260 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-003-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.