Orders LinkContactUsLinkComponent
Hydroquinone monomethyl ether 98+%
Bottle, 250 g
  AAA14845-30
 :  A14845-30
Hydroquinone monomethyl ether 98+%
  AAA14845-30
 :  150-76-5
Hydroquinone monomethyl ether 98+%
Hydroquinone monomethyl ether 98+%
Bottle, 250 g
 :  150-76-5
$66.16
Each (250g)
Purity: 98+% CAS No.: 150-76-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 132°(269°F) Melting Point: 55-58° Boiling Point: 243-246° Beilstein Reference: 507924 MDL No.: MFCD00002332
 :  p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole
  •  :150-76-5
  •  :C₇H₈O₂
  •  :MFCD00002332
  •  :124.14 g/mol
  •  :243…246 °C
  •  :55…58 °C
  •  :1.550
  •  :132 °C (269 °F)
  •  :132 °C(269 °F)
  •  :Ambient
  •  :205-769-8
  • Pack type:
    Bottle
  • Size:
    250 g