Order Entry
United States
Orders LinkContactUsLinkComponent
 

Methyl violet 2B

Bottle, 250 g
  AAA12159-30
 
 
 :  A12159-30
Methyl violet 2B
  AAA12159-30
 :  8004-87-3
Methyl violet 2B
Methyl violet 2B

Bottle, 250 g

 :  8004-87-3
 
$152.66
   
Each (250g)
 
   
 

MDL: MFCD00012568 Notes: Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)

CAS No.: 8004-87-3 Molecular Formula: C24H27N3.HCl Formula Weight: 393.96 Odor: Odorless MDL No.: MFCD00012568

 :  Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
  •  :8004-87-3
  •  :C₂₄H₂₈ClN₃
  •  :MFCD00012568
  •  :393.96 g/mol
  •  :137 °C
  •  :Ambient
  •  :UN3077
  •  :9,III
 

 

  • Pack type:
    Bottle
  • Size:
    250 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.