Order Entry
United States
Orders LinkContactUsLinkComponent
Hardness 1 Buffer Solution, Hach

Hardness 1 Buffer Solution, Hach

Hardness 1 Buffer Solution, 100 ml, EDTA, Marked dropping bottle (MDB)
Catalog # 97026-364
Product available on GSA Advantage®
Hardness 1 Buffer Solution, Hach
Catalog # 97026-364
Supplier:  
Product available on GSA Advantage®
CAS Number:  
Hardness 1 Buffer Solution, Hach
Hardness 1 Buffer Solution, Hach

Hardness 1 Buffer Solution, 100 ml, EDTA, Marked dropping bottle (MDB)

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SOLUTION BUFFER 100ML Catalog #104086-790

 

WATER HARDNESS BUFFER 500ML Catalog #101226-388

 

Product Details & Documents

Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C

Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.

 

Specifications

Specifications

  • Chemical method:
    EDTA
  • Pack type:
    Marked dropping bottle (MDB)
  • Description:
    Hardness 1 Buffer Solution
  • Size:
    100 ml
  • Cat. No.:
    97026-364
  • Method:
    8226, 8213; EDTA Titration

Specifications