Order Entry
United States
Orders LinkContactUsLinkComponent
PermaRed/AP, Diagnostic Biosystems

PermaRed/AP, Diagnostic Biosystems

PermaRed/AP, 30 ml
Catalog # 95041-484
Supplier #:  K049
PermaRed/AP, Diagnostic Biosystems
Catalog # 95041-484
CAS Number:  
PermaRed/AP, Diagnostic Biosystems
PermaRed/AP, Diagnostic Biosystems

PermaRed/AP, 30 ml

CAS Number:  
Price
$138.71
Log in for availability
Each (30ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PermaRed/AP is a substrate chromogen system for use with alkaline phosphatase (AP) detection in immunohistochemistry (IHC) or in situ hybridizations (ISH).

PermaRed/AP produces a brilliant dark red color and insoluble in organic solvents; therefore sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted.

Documents

Specifications

Specifications

  • Description:
    PermaRed/AP
  • Size:
    30 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.