Order Entry
United States
Orders LinkContactUsLinkComponent
 

Collodion USP

Glass, amber, 4 L
Catalog # 95030-696
Flammable
Irritant
 :  CO120-4LTGL
Collodion USP
Catalog # 95030-696
CAS Number:  9004-70-0
Collodion USP

Glass, amber, 4 L

CAS Number:  9004-70-0
Price
$1,117.41
Log in for availability
Each (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

  • CAS Number:9004-70-0
  • MDL Number:MFCD00081525
  • Merck Index:13,02508
  • Boiling Pt:∼34 °C (1013 hPa)
  • Melting Pt:160 °C
  • Density:0.75 g/cm³ (20 °C)
  • Flash Pt:–19 °C
  • Storage Temperature:Ambient
  • UN:2059
  • ADR:3,II
Specifications

Specification Test Results

  • Assay:
    5% Min.

Specifications

  • Pack type:
    Glass, amber
  • Size:
    4 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.