Order Entry
United States
Orders LinkContactUsLinkComponent
Cell-Chex Controls, Streck

Cell-Chex Controls, Streck

Levels 1 and 2, 4 each×2.0 ml
Catalog # 87005-248
Supplier:   Streck Laboratories
Supplier #:  212420
Cell-Chex Controls, Streck
Catalog # 87005-248
CAS Number:  
Cell-Chex Controls, Streck
Cell-Chex Controls, Streck

Levels 1 and 2, 4 each×2.0 ml

CAS Number:  
Price
$307.60
Log in for availability
Box of 4
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell-Chex controls offer spinal and body fluid procedural control for red blood cell and white blood cell counts, crystal identification and white blood cell differentiation

Level 1-UC contains urate crystals, and Level 1-CC contains CPPD crystals to aid in identification of crystals in synovial fluid. The white blood cells can be differentiated when stained in the same manner as a patient sample. The two-level control is assayed for manual cell counting, five-part white blood cell differential and two-part polymorphonuclear and mononuclear differential

Ordering information: Cell-Chex controls maintain a closed-vial stability for 6 months and an open-vial stability for 30 days when stored at 2 to 10°C (36 to 50°F).

Specifications

Specifications

  • Description:
    Levels 1 and 2
  • Size:
    4 each×2.0 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.