Orders LinkContactUsLinkComponent
Histosette® II Tissue Processing/Embedding Cassettes, Simport
Gray
  14230-484
 :  M485-9
Histosette® II Tissue Processing/Embedding Cassettes, Simport
  14230-484
Histosette® II Tissue Processing/Embedding Cassettes, Simport
Histosette® II Tissue Processing/Embedding Cassettes, Simport
Gray
$288.39
Case of 1,000
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. Anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments. Colors are non-cytotoxic and nonmetallic
 : Lids are supplied unattached. Packed in two dispenser boxes of 500 cassettes and one dispenser box of 1000 lids.
  • Color:
    Gray