$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
EPH FRACTIONATION CHECK SOLN
ULSWA-330-1
:
Ultra Scientific
EPH FRACTIONATION CHECK SOLN
ULSWA-330-1
:
Ultra Scientific
:
SWA-330-1
ghs_SWA-330-1.pdf
VWR GHS Compliance Letter (US)
Frequently Bought Together
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
: Kimberly-Clark
Clorox® Concentrated Germicidal Bleach
: Janitorial Supplies
VWR® Disposable Serological Pipettes
: VWR International
VWR® Broken Glass Disposal Cartons
: VWR International
VWR® Lab Markers
: VWR International
VWR® Powder-Free Nitrile Examination Gloves
: VWR International
VWR® Disposable Petri Dishes
: VWR International
PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark
: Ansell Healthcare
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
: VWR International
VWR® Nitrile Examination Gloves, Black
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International