1,2,6-Hexanetriol ≥96.0%
Product Details & Documents
CAS Number: 106-69-4 MDL Number: MFCD00002976 Molecular Formula: C6H14O3 Molecular Weight: 134.18 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 178 Flash Point (°C): 191 Specific Gravity (20/20): 1.10
Product Categories: Bioscience, Cosmetic Ingredients for Research and Experimental Use Other Product Names / Synonyms: 1,2,6-Trihydroxyhexane
- CAS Number:106-69-4
- Formula:C₆H₁₄O₃
- MDL Number:MFCD00002976
- Molecular Weight:134.18 g/mol
- Boiling Pt:178 °C
- Melting Pt:25…32 °C
- Density:1.10 (20 °C)
- Flash Pt:191 °C
- Storage Temperature:Ambient
- EINECS:203-424-6
Documents
Certificates
Search by Lot NumberTCI America Production Chemicals.pdf
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specification Test Results
- Purity/Analysis Method:96.0% (GC)
Specifications
- Size:25 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



