2-Propanol ≥70%, Supelco®
Poly bottle, 4 L
Supplier: MilliporeSigma
: PX1840-4
Jump to:
Product Details & Documents
Reagent
Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Density:0.85 g/cm3 (20 °C)
- Flash Pt:12
- Storage Temperature:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
Specifications
Specification Test Results
- Color (APHA):10APHA max
- Expiration date:36 months from mfg date
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Specific Gravity at 20/20°C:0.872-0.883
Specifications
- Pack type:Poly bottle
- Size:4 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



