Order Entry
United States
Orders LinkContactUsLinkComponent
(±)-2-Hexanol 99%

(±)-2-Hexanol 99%

bottle, 100 g
Catalog # AAAL05993-22
Flammable
Irritant
Supplier #:  L05993-22
(±)-2-Hexanol 99%
Catalog # AAAL05993-22
CAS Number:  626-93-7
(±)-2-Hexanol 99%
(±)-2-Hexanol 99%

bottle, 100 g

CAS Number:  626-93-7
Price
$206.24
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 626-93-7 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 45°(113°F) Refractive Index: 1.4140 Odor: Characteristic Melting Point: -23° Boiling Point: 137-139° Beilstein Reference: 1718996 UN No.: 2282 MDL No.: MFCD00004585

Synonyms:  (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
  • CAS Number:626-93-7
  • Formula:C₆H₁₄O
  • MDL Number:MFCD00004585
  • Molecular Weight:102.18 g/mol
  • Boiling Pt:137…139 °C
  • Melting Pt:–23 °C
  • Density:0.810
  • Flash Pt:45 °C (113 °F)
  • Storage Temperature:Ambient
  • EINECS:210-971-4
  • UN:2282
  • ADR:3,III

Documents

Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.