Order Entry
United States
Orders LinkContactUsLinkComponent
Dapsone 98%

Dapsone 98%

Bottle, 100 g
Catalog # AAA18976-22
Irritant
Supplier #:  A18976-22
Dapsone 98%
Catalog # AAA18976-22
CAS Number:  80-08-0
Dapsone 98%
Dapsone 98%

Bottle, 100 g

CAS Number:  80-08-0
Price
$53.73
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98% CAS No.: 80-08-0 Molecular Formula: C12H12N2O2S Formula Weight: 248.30 Odor: Odorless Melting Point: 175-178° Beilstein Reference: 788055 Merck Reference: 14,2822 MDL No.: MFCD00007887

Synonyms:  4-Aminophenyl sulfone, 1-Amino-4-((4-aminophenyl)sulfonyl)benzene, 4,4'-sulfonyldianiline, 4,4'-Sulfonyldianiline, 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Diamino diphenyl sulfone, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone, 4-[(4-Aminophenyl)sulfonyl]aniline
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Merck Index:14,2822
  • Melting Pt:175…178 °C
  • Storage Temperature:Ambient
  • EINECS:201-248-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.