n-Decane 99%
Product Details & Documents
MDL: MFCD00008954 Beilstein: 1696981 Soluble in alcohol and ethers
Purity: 99% CAS No.: 124-18-5 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.730 Refractive Index: 1.4115 Appearance: Colorless Odor: Petrol-like Melting Point: -30° Boiling Point: 172-174° Solubility: Soluble in alcohol and ethers Beilstein Reference: 1696981 UN No.: 2247 MDL No.: MFCD00008954
- CAS Number:124-18-5
- Formula:H₃C(CH₂)₈CH₃
- MDL Number:MFCD00008954
- Molecular Weight:142.28 g/mol
- Boiling Pt:172…174 °C
- Melting Pt:–30 °C
- Density:0.730
- Flash Pt:46 °C (115 °F)
- Storage Temperature:Ambient
- EINECS:204-686-4
- UN:2247
- ADR:3,III
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Pack type:Bottle
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



