D-(-)-Mannitol ≥99.0% ACS
Bottle, 500 g
Supplier: Thermo Scientific Chemicals
: 033342-36
D-(-)-Mannitol ≥99.0% ACS
Catalog # AA33342-36
Supplier: Thermo Scientific Chemicals
CAS Number: 69-65-8
Jump to:
Product Details & Documents
MDL: MFCD00064287 Beilstein Registry No.: 1721898 Soluble in water, alcohol, pyridine, aniline. Insoluble in ether
CAS No.: 69-65-8 Molecular Formula: HOCH2(CHOH)4CH2OH Formula Weight: 182.17 Physical Form: Crystalline Density: 1.489 Appearance: White Odor: Odorless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Solubility: Soluble in water, alcohol, pyridine, aniline. Insoluble in ether Beilstein Reference: 1721898 Merck Reference: 14,5745 MDL No.: MFCD00064287
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:14,5745
- Boiling Pt:290…295 °C (3.5 mmHg)
- Melting Pt:165…167 °C
- Density:1.489
- Storage Temperature:Ambient
- EINECS:200-711-8
Specifications
Specification Test Results
- Insoluble matter:0.01%
- Residue after ignition:0.01%
- Titratable acid:0.0008meq/g
- Reducing sugars:Pass test
- Heavy Metals (as Pb):5 ppm
- Specific rotation at 25°:+23.3° to +24.3°
Specifications
- Pack type:Bottle
- Size:500 g



