Order Entry
United States
Orders LinkContactUsLinkComponent
Sodium nitrite ≥97% ACS

Sodium nitrite ≥97% ACS

Bottle, 25 g
  AA14244-14
 
 
 
Supplier #:  014244-14
Sodium nitrite ≥97% ACS
  AA14244-14
 :  7632-00-0
Sodium nitrite ≥97% ACS

Bottle, 25 g

 :  7632-00-0
 
$51.64
   
Each (25g)
 
   
 

MDL: MFCD00011118 Notes: Decomposes above 320°C Soluble in water Fieser: 1,1097 11,491 13,282 16,308 18,336 19,313 20,350 21,405

Purity: 97% min CAS No.: 7632-00-0 Molecular Formula: NaNO2 Formula Weight: 69.00 Physical Form: Granular Density: 2.17 Melting Point: 271° Solubility: Soluble in water Merck Reference: 14,8648 UN No.: 1500 MDL No.: MFCD00011118

 :  Diazotizing salts
  •  :7632-00-0
  •  :NaNO₂
  •  :MFCD00011118
  •  :69 g/mol
  •  :14,8648
  •  :320 °C (1013 hPa)
  •  :271 °C
  •  :2.17
  •  :Ambient
  •  :231-555-9
  •  :1500
  •  :5.1,III

Documents

 

 

  • Insoluble matter:
    0.01%
  • Cl:
    0.005%
  • SO3:
    0.01%
  • Ca:
    0.01%
  • Heavy Metals (as Pb):
    0.001%
  • Fe:
    0.001%
  • K:
    0.005%

 

  • Pack type:
    Bottle
  • Size:
    25 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.