Order Entry
United States
Orders LinkContactUsLinkComponent
Leupeptin

Leupeptin

5 mg
Catalog # 97063-924
Supplier:   VWR
Supplier #:  M180-5MG
Leupeptin
Catalog # 97063-924
Supplier:  VWR
CAS Number:  
Leupeptin
Leupeptin

5 mg

CAS Number:  
Price
$262.09
Log in for availability
Each (5mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Product is animal-free

  • Activity: Working range: 0.5 µg/ml (1 µM)

Extracted proteins are exposed to the threat of degradation by proteases from the moment cells or tissues are lysed. Proteases present in the living cell are released from their regulated environment into the extract where they may freely degrade their targets, resulting in drastically reduced protein yield and sample
quality. Complex protein solutions are typically treated with a mixture of several protease inhibitors in order to preserve the native state of the proteins.

A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin,papain, kallikrein, thrombin and cathepsin A and B.

Specifications

Specifications

  • Size:
    5 mg
  • Cat. No.:
    97063-924
  • Protein/Peptide Name:
    Leupeptin
  • Grade:
    Proteomics Grade

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.