Order Entry
United States
Orders LinkContactUsLinkComponent
 

VWR® Phosphate Buffer Solution

500 mL
Catalog # 97062-960
VWR® Phosphate Buffer Solution
Catalog # 97062-960
Supplier:  
CAS Number:  
VWR® Phosphate Buffer Solution
VWR® Phosphate Buffer Solution

500 mL

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER BLOCKING PBST FIRST CHOICE 500ML Catalog #89167-872

Product 89167-872 is a replacement for 97062-960 and has the same packaging and specifications, but may be a different quantity.

PBS-POLYSORBATE PH 7.5 Catalog #97062-958

Product 97062-958 is a replacement for 97062-960 and has the same packaging and specifications, but may be a different quantity.

PHOSPHATE BUFFERED SALINE SOLUTION 500ML Catalog #76014-826

Product 76014-826 is a replacement for 97062-960 and has the same packaging and specifications, but may be a different quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, pH 7.5 is a ready-to-use 1X concentration

Specifications

Specifications

  • Size:
    500 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.