Order Entry
United States
Orders LinkContactUsLinkComponent
 

Phosphate Buffer, BD Diagnostics

Phosphate Buffer, 300 ml
Catalog # 95062-840
Phosphate Buffer, BD Diagnostics
Catalog # 95062-840
Supplier:  
CAS Number:  
Phosphate Buffer, BD Diagnostics

Phosphate Buffer, 300 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PHOSPHATE BUFFER PH 7.2 1L Catalog #BDH7345-1

 

Product Details & Documents
Specifications

Specifications

  • Description:
    Phosphate Buffer
  • Volume:
    300 ml

Specifications